Drukuj

Anti-POLE Rabbit Monoclonal Antibody [clone: EPR28640-56]

Dostawca: Abcam
AB318136-20UL AB318136-100UL AB318136-1ML
ABCAAB318136-20ULEA 1090 PLN
ABCAAB318136-20UL ABCAAB318136-100UL ABCAAB318136-1ML
Anti-POLE Rabbit Monoclonal Antibody [clone: EPR28640-56]
Przeciwciała
Rabbit monoclonal [EPR28640-56] to POLE (mutated P286R).

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: POLE
Clonality: Monoclonal
Clone: EPR28640-56
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR