Drukuj

Anti-Netrin G1 Rabbit Monoclonal Antibody [clone: EPR27984-506]

Dostawca: Abcam
AB319038-100UG AB319038-1MG
ABCAAB319038-100UGEA 4530 PLN
ABCAAB319038-100UG ABCAAB319038-1MG
Anti-Netrin G1 Rabbit Monoclonal Antibody [clone: EPR27984-506]
Przeciwciała
Rabbit monoclonal [EPR27984-506] to Netrin G1 - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Netrin G1
Clonality: Monoclonal
Clone: EPR27984-506
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR