Drukuj

Anti-MAP2 Chicken Monoclonal Antibody [clone: EPR19691]

Dostawca: Abcam
AB319979-20UL AB319979-100UL AB319979-1ML
ABCAAB319979-20ULEA 1170 PLN
ABCAAB319979-20UL ABCAAB319979-100UL ABCAAB319979-1ML
Anti-MAP2 Chicken Monoclonal Antibody [clone: EPR19691]
Przeciwciała
Chicken monoclonal [EPR19691] to MAP2 - Chimeric.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone: EPR19691
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR