Drukuj

Anti-Perilipin 3/TIP47 Rabbit Monoclonal Antibody [clone: EPR28801-93]

Dostawca: Abcam
AB315476-20UL AB315476-100UL AB315476-1ML
ABCAAB315476-20ULEA 1090 PLN
ABCAAB315476-20UL ABCAAB315476-100UL ABCAAB315476-1ML
Anti-Perilipin 3/TIP47 Rabbit Monoclonal Antibody [clone: EPR28801-93]
Przeciwciała
Rabbit monoclonal [EPR28801-93] to Perilipin 3/TIP47.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Perilipin 3/TIP47
Clonality: Monoclonal
Clone: EPR28801-93
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR