Drukuj

Anti-ENPP2/ATX Rabbit Monoclonal Antibody [clone: EPR25942-70]

Dostawca: Abcam
AB315984-1MG AB315984-100UG
ABCAAB315984-1MGEA 40850 PLN
ABCAAB315984-1MG ABCAAB315984-100UG
Anti-ENPP2/ATX Rabbit Monoclonal Antibody [clone: EPR25942-70]
Przeciwciała
Rabbit monoclonal [EPR25942-70] to ENPP2/ATX - BSA and Azide free (Capture).

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ENPP2/ATX
Clonality: Monoclonal
Clone: EPR25942-70
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR