Drukuj

Anti-Ki67 Chicken Monoclonal Antibody [clone: SP6]

Dostawca: Abcam
AB320709-20UL AB320709-100UL AB320709-1ML
ABCAAB320709-20ULEA 1170 PLN
ABCAAB320709-20UL ABCAAB320709-100UL ABCAAB320709-1ML
Anti-Ki67 Chicken Monoclonal Antibody [clone: SP6]
Przeciwciała
Chicken monoclonal [SP6] to Ki67 - Chimeric.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Ki67
Clonality: Monoclonal
Clone: SP6
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human;Rat;Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR