Drukuj

Anti-Colec12 Rabbit Monoclonal Antibody

Dostawca: Abcam
AB317630-20UL AB317630-100UL AB317630-1ML
ABCAAB317630-20ULEA 1090 PLN
ABCAAB317630-20UL ABCAAB317630-100UL ABCAAB317630-1ML
Anti-Colec12 Rabbit Monoclonal Antibody
Przeciwciała
Rabbit monoclonal [EPR29371-589] to Colec12.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Colec12
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR