Drukuj
Anti-AP4B1 Rabbit Monoclonal Antibody [clone: EPR28048-85]
Dostawca: Abcam
AB316003-1MG AB316003-100UG
ABCAAB316003-1MGEA 40660 PLN
ABCAAB316003-1MG ABCAAB316003-100UG
Anti-AP4B1 Rabbit Monoclonal Antibody [clone: EPR28048-85]
Przeciwciała
Rabbit monoclonal [EPR28048-85] to AP4B1 - BSA and Azide free.
Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: AP4B1
Clonality: Monoclonal
Clone: EPR28048-85
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat