Drukuj

Anti-Annexin V/ANXA5 Rabbit Monoclonal Antibody [clone: EPR28047-61] (Alexa Fluor® 647)

Dostawca: Abcam
AB318176-10UL AB318176-100UL
ABCAAB318176-10ULEA 1060 PLN
ABCAAB318176-10UL ABCAAB318176-100UL
Anti-Annexin V/ANXA5 Rabbit Monoclonal Antibody [clone: EPR28047-61] (Alexa Fluor® 647)
Przeciwciała
Alexa Fluor® 647 Rabbit monoclonal [EPR28047-61] to Annexin V/ANXA5.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Annexin V/ANXA5
Clonality: Monoclonal
Clone: EPR28047-61
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR