Drukuj

Anti-HDAC1+HDAC2+HDAC3+HDAC8 Rabbit Recombinant Multiclonal Antibody [clone: RM1172]

Dostawca: Abcam
AB319059-20UL AB319059-100UL AB319059-1ML
ABCAAB319059-20ULEA 916 PLN
ABCAAB319059-20UL ABCAAB319059-100UL ABCAAB319059-1ML
Anti-HDAC1+HDAC2+HDAC3+HDAC8 Rabbit Recombinant Multiclonal Antibody [clone: RM1172]
Przeciwciała
Rabbit recombinant multiclonal [RM1172] to HDAC1 + HDAC2 + HDAC3 + HDAC8.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HDAC1+HDAC2+HDAC3+HDAC8
Clonality: Recombinant Multiclonal
Clone: RM1172
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR