Drukuj

Anti-bat SARS-like coronavirus RsSHC014 Spike glycoprotein Human Monoclonal Antibody [clone: abd166]

Dostawca: Abcam
AB317329-100UG AB317329-20UG AB317329-1MG
ABCAAB317329-100UGEA 3090 PLN
ABCAAB317329-100UG ABCAAB317329-20UG ABCAAB317329-1MG
Anti-bat SARS-like coronavirus RsSHC014 Spike glycoprotein Human Monoclonal Antibody [clone: abd166]
Przeciwciała
Human monoclonal [abd166] to bat SARS-like coronavirus RsSHC014 Spike glycoprotein.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: bat SARS-like coronavirus RsSHC014 Spike glycoprotein
Clonality: Monoclonal
Clone: abd166
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR