Drukuj
Anti-ADAM22 Rabbit Monoclonal Antibody [clone: EPR28037-56]
Dostawca: Abcam
AB314443-1MG AB314443-100UG
ABCAAB314443-1MGEA 40660 PLN
ABCAAB314443-1MG ABCAAB314443-100UG
Anti-ADAM22 Rabbit Monoclonal Antibody [clone: EPR28037-56]
Przeciwciała
Rabbit monoclonal [EPR28037-56] to ADAM22 - BSA and Azide free.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ADAM22
Clonality: Monoclonal
Clone: EPR28037-56
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat