Drukuj
Anti-Clostridium difficile Toxin B Rabbit Monoclonal Antibody [clone: EPR23357-19]
Dostawca: Abcam
AB315113-1MG AB315113-100UG
ABCAAB315113-1MGEA 40850 PLN
ABCAAB315113-1MG ABCAAB315113-100UG
Anti-Clostridium difficile Toxin B Rabbit Monoclonal Antibody [clone: EPR23357-19]
Przeciwciała
Rabbit monoclonal [EPR23357-19] to Clostridium difficile Toxin B - BSA and Azide free (Capture).
Type: Primary
Antigen: Clostridium difficile Toxin B
Clonality: Monoclonal
Clone: EPR23357-19
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: C dif