Drukuj

Anti-Clostridium difficile Toxins A + B Rabbit Monoclonal Antibody [clone: EPR23358-169]

Dostawca: Abcam
AB315117-1MG AB315117-100UG
ABCAAB315117-1MGEA 40850 PLN
ABCAAB315117-1MG ABCAAB315117-100UG
Anti-Clostridium difficile Toxins A + B Rabbit Monoclonal Antibody [clone: EPR23358-169]
Przeciwciała
Rabbit monoclonal [EPR23358-169] to Clostridium difficile Toxins A + B - BSA and Azide free (Detector).

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Clostridium difficile Toxins A + B
Clonality: Monoclonal
Clone: EPR23358-169
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Clostridium difficile
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR