Drukuj

Anti-Tyrosine Hydroxylase Mouse Monoclonal Antibody [clone: EP1532Y]

Dostawca: Abcam
AB317786-20UL AB317786-100UL AB317786-1ML
ABCAAB317786-20ULEA 1170 PLN
ABCAAB317786-20UL ABCAAB317786-100UL ABCAAB317786-1ML
Anti-Tyrosine Hydroxylase Mouse Monoclonal Antibody [clone: EP1532Y]
Przeciwciała
Mouse monoclonal [EP1532Y] to Tyrosine Hydroxylase - Chimeric.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Monoclonal
Clone: EP1532Y
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR