Drukuj
Anti-Pseudomonas Outer membrane porin F Rabbit Monoclonal Antibody [clone: EPR28629-91]
Dostawca: Abcam
AB316974-100UG AB316974-1MG
ABCAAB316974-100UGEA 4530 PLN
ABCAAB316974-100UG ABCAAB316974-1MG
Anti-Pseudomonas Outer membrane porin F Rabbit Monoclonal Antibody [clone: EPR28629-91]
Przeciwciała
Rabbit monoclonal [EPR28629-91] to Pseudomonas Outer membrane porin F - BSA and Azide free.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Pseudomonas Outer membrane porin F
Clonality: Monoclonal
Clone: EPR28629-91
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: P aer