Drukuj
Anti-epithelial Sodium Channel alpha Rabbit Monoclonal Antibody [clone: EPR28997-46]
Dostawca: Abcam
AB322260-20UL AB322260-100UL AB322260-1ML
ABCAAB322260-20ULEA 1090 PLN
ABCAAB322260-20UL ABCAAB322260-100UL ABCAAB322260-1ML
Anti-epithelial Sodium Channel alpha Rabbit Monoclonal Antibody [clone: EPR28997-46]
Przeciwciała
Rabbit monoclonal [EPR28997-46] to epithelial Sodium Channel alpha.
Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: epithelial Sodium Channel alpha
Clonality: Monoclonal
Clone: EPR28997-46
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human