Drukuj

Anti-Nogo A+Nogo D Rabbit Monoclonal Antibody [clone: EPR26286-56]

Dostawca: Abcam
AB315792-20UL AB315792-100UL AB315792-1ML
ABCAAB315792-20ULEA 1090 PLN
ABCAAB315792-20UL ABCAAB315792-100UL ABCAAB315792-1ML
Anti-Nogo A+Nogo D Rabbit Monoclonal Antibody [clone: EPR26286-56]
Przeciwciała
Rabbit monoclonal [EPR26286-56] to Nogo A + Nogo D.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Nogo A+Nogo D
Clonality: Monoclonal
Clone: EPR26286-56
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR