Drukuj

Anti-Lactate Dehydrogenase A (LDHA) Rabbit Monoclonal Antibody [clone: EPR25915-6]

Dostawca: Abcam
AB317640-100UG AB317640-1MG
ABCAAB317640-100UGEA 4080 PLN
ABCAAB317640-100UG ABCAAB317640-1MG
Anti-Lactate Dehydrogenase A (LDHA) Rabbit Monoclonal Antibody [clone: EPR25915-6]
Przeciwciała
Rabbit monoclonal [EPR25915-6] to Lactate Dehydrogenase A (LDHA) - BSA and Azide free (Capture).

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Lactate Dehydrogenase A (LDHA)
Clonality: Monoclonal
Clone: EPR25915-6
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR