Drukuj
Anti-Plexin B1 Rabbit Monoclonal Antibody [clone: EPR23781-82]
Dostawca: Abcam
AB281260-1 AB281260-100
ABCAAB281260-1EA 38520 PLN
ABCAAB281260-1 ABCAAB281260-100
Anti-Plexin B1 Rabbit Monoclonal Antibody [clone: EPR23781-82]
Przeciwciała
Rabbit monoclonal [EPR23781-82] to Plexin B1 - BSA and Azide free (Capture).
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Plexin B1
Clonality: Monoclonal
Clone: EPR23781-82
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human