Drukuj

Anti-KIF5C Rabbit Monoclonal Antibody [clone: EPR16224-32]

Dostawca: Abcam
AB193352-100 AB193352-10 AB193352-1ML
ABCAAB193352-100EA 3030 PLN
ABCAAB193352-100 ABCAAB193352-10 ABCAAB193352-1ML
Anti-KIF5C Rabbit Monoclonal Antibody [clone: EPR16224-32]
Przeciwciała
Rabbit monoclonal [EPR16224-32] to KIF5C.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: KIF5C
Clonality: Monoclonal
Clone: EPR16224-32
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR