Drukuj

Anti-M Cadherin Rabbit Monoclonal Antibody [clone: EPR25401-56]

Dostawca: Abcam
AB309477-100UL AB309477-20UL AB309477-1ML
ABCAAB309477-100ULEA 3090 PLN
ABCAAB309477-100UL ABCAAB309477-20UL ABCAAB309477-1ML
Anti-M Cadherin Rabbit Monoclonal Antibody [clone: EPR25401-56]
Przeciwciała
Rabbit monoclonal [EPR25401-56] to M Cadherin.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: M Cadherin
Clonality: Monoclonal
Clone: EPR25401-56
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR