Drukuj

Anti-F4/80 Rabbit Monoclonal Antibody [clone: EPR20548]

Dostawca: Abcam
AB213200-100 AB213200-20UL AB213200-1ML
ABCAAB213200-100EA 2110 PLN
ABCAAB213200-100 ABCAAB213200-20UL ABCAAB213200-1ML
Anti-F4/80 Rabbit Monoclonal Antibody [clone: EPR20548]
Przeciwciała
Rabbit monoclonal [EPR20548] to F4/80.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: F4/80
Clonality: Monoclonal
Clone: EPR20548
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR