Drukuj

Anti-HLA-DR Mouse Monoclonal Antibody [clone: TAL 1B5] (Alexa Fluor® 555)

Dostawca: Abcam
AB309506-10UL AB309506-100UL
ABCAAB309506-10ULEA 1060 PLN
ABCAAB309506-10UL ABCAAB309506-100UL
Anti-HLA-DR Mouse Monoclonal Antibody [clone: TAL 1B5] (Alexa Fluor® 555)
Przeciwciała
Alexa Fluor® 555 Mouse monoclonal [TAL 1B5] to HLA-DR.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: TAL 1B5
Conjugation: Alexa Fluor® 555
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR