Drukuj

Anti-Claudin 1 Rabbit Monoclonal Antibody [clone: EPR9306]

Dostawca: Abcam
AB180158-100 AB180158-1ML
ABCAAB180158-100EA 3420 PLN
ABCAAB180158-100 ABCAAB180158-1ML
Anti-Claudin 1 Rabbit Monoclonal Antibody [clone: EPR9306]
Przeciwciała
Rabbit monoclonal [EPR9306] to Claudin 1.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Claudin 1
Clonality: Monoclonal
Clone: EPR9306
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR