Drukuj
Anti-Lamin A+Lamin B1+Lamin C Rabbit Monoclonal Antibody [clone: EPR4068]
Dostawca: Abcam
AB226043-1 AB226043-100
ABCAAB226043-1EA 40660 PLN
ABCAAB226043-1 ABCAAB226043-100
Anti-Lamin A+Lamin B1+Lamin C Rabbit Monoclonal Antibody [clone: EPR4068]
Przeciwciała
Rabbit monoclonal [EPR4068] to Lamin A + Lamin B1 + Lamin C - BSA and Azide free.
Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Lamin A+Lamin B1+Lamin C
Clonality: Monoclonal
Clone: EPR4068
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human