Drukuj

Anti-Neurofilament heavy polypeptide Rabbit Monoclonal Antibody [clone: EPR20020]

Dostawca: Abcam
AB223139-100 AB223139-1
ABCAAB223139-100EA 4530 PLN
ABCAAB223139-100 ABCAAB223139-1
Anti-Neurofilament heavy polypeptide Rabbit Monoclonal Antibody [clone: EPR20020]
Przeciwciała
Rabbit monoclonal [EPR20020] to Neurofilament heavy polypeptide - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Neurofilament heavy polypeptide
Clonality: Monoclonal
Clone: EPR20020
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR