Drukuj
Anti-LIX Rabbit Monoclonal Antibody [clone: EPR22392-51]
Dostawca: Abcam
AB267436-100 AB267436-1
ABCAAB267436-100EA 4170 PLN
ABCAAB267436-100 ABCAAB267436-1
Anti-LIX Rabbit Monoclonal Antibody [clone: EPR22392-51]
Przeciwciała
Rabbit monoclonal [EPR22392-51] to LIX - BSA and Azide free (Detector).
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: LIX
Clonality: Monoclonal
Clone: EPR22392-51
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse