Drukuj

Anti-CD33 Rabbit Monoclonal Antibody [clone: EPR24370-124]

Dostawca: Abcam
AB270942-100 AB270942-20UL AB270942-1ML
ABCAAB270942-100EA 3170 PLN
ABCAAB270942-100 ABCAAB270942-20UL ABCAAB270942-1ML
Anti-CD33 Rabbit Monoclonal Antibody [clone: EPR24370-124]
Przeciwciała
Rabbit monoclonal [EPR24370-124] to CD33.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: EPR24370-124
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR