Drukuj

Anti-SAP97 Rabbit Monoclonal Antibody [clone: EPR26467-123]

Dostawca: Abcam
AB300481-10 AB300481-100 AB300481-20UL AB300481-1ML
ABCAAB300481-10EA 1030 PLN
ABCAAB300481-10 ABCAAB300481-100 ABCAAB300481-20UL ABCAAB300481-1ML
Anti-SAP97 Rabbit Monoclonal Antibody [clone: EPR26467-123]
Przeciwciała
Rabbit monoclonal [EPR26467-123] to SAP97.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SAP97
Clonality: Monoclonal
Clone: EPR26467-123
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR