Drukuj
Anti-DDX4 / MVH Rabbit Monoclonal Antibody [clone: EPR24148-58]
Dostawca: Abcam
AB270534-100 AB270534-20UL AB270534-1ML
ABCAAB270534-100EA 3170 PLN
ABCAAB270534-100 ABCAAB270534-20UL ABCAAB270534-1ML
Anti-DDX4 / MVH Rabbit Monoclonal Antibody [clone: EPR24148-58]
Rabbit monoclonal [EPR24148-58] to DDX4 / MVH.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: DDX4 / MVH
Clonality: Monoclonal
Clone: EPR24148-58
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat