Drukuj

Anti-pro Caspase-1 + p10 + p12 Rabbit Monoclonal Antibody [clone: EPR16883]

Dostawca: Abcam
AB179515-100 AB179515-1ML
ABCAAB179515-100EA 3250 PLN
ABCAAB179515-100 ABCAAB179515-1ML
Anti-pro Caspase-1 + p10 + p12 Rabbit Monoclonal Antibody [clone: EPR16883]
Przeciwciała
Rabbit monoclonal [EPR16883] to pro Caspase-1 + p10 + p12.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: pro Caspase-1 + p10 + p12
Clonality: Monoclonal
Clone: EPR16883
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR