Drukuj

Anti-Proteasome 20S LMP2 Rabbit Monoclonal Antibody [clone: EPR13785]

Dostawca: Abcam
AB184172-100 AB184172-10 AB184172-1ML
ABCAAB184172-100EA 3030 PLN
ABCAAB184172-100 ABCAAB184172-10 ABCAAB184172-1ML
Anti-Proteasome 20S LMP2 Rabbit Monoclonal Antibody [clone: EPR13785]
Przeciwciała
Rabbit monoclonal [EPR13785] to Proteasome 20S LMP2.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Proteasome 20S LMP2
Clonality: Monoclonal
Clone: EPR13785
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR