Drukuj
Anti-Collagen II Rabbit Monoclonal Antibody [clone: EPR12268]
Dostawca: Abcam
AB239007-100 AB239007-1
ABCAAB239007-100EA 4530 PLN
ABCAAB239007-100 ABCAAB239007-1
Anti-Collagen II Rabbit Monoclonal Antibody [clone: EPR12268]
Przeciwciała
Rabbit monoclonal [EPR12268] to Collagen II - BSA and Azide free.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Collagen II
Clonality: Monoclonal
Clone: EPR12268
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat;Human