Drukuj

Anti-CTHRC1 Rabbit Monoclonal Antibody [clone: EPR22851-144]

Dostawca: Abcam
AB274446-100 AB274446-1
ABCAAB274446-100EA 3860 PLN
ABCAAB274446-100 ABCAAB274446-1
Anti-CTHRC1 Rabbit Monoclonal Antibody [clone: EPR22851-144]
Przeciwciała
Rabbit monoclonal [EPR22851-144] to CTHRC1 - BSA and Azide free (Capture).

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CTHRC1
Clonality: Monoclonal
Clone: EPR22851-144
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR