Drukuj

Anti-F4/80 Rabbit Monoclonal Antibody [clone: EPR26545-166]

Dostawca: Abcam
AB300421-20UL AB300421-1ML
ABCAAB300421-20ULEA 1110 PLN
ABCAAB300421-20UL ABCAAB300421-1ML
Anti-F4/80 Rabbit Monoclonal Antibody [clone: EPR26545-166]
Przeciwciała
Rabbit monoclonal [EPR26545-166] to F4/80.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: F4/80
Clonality: Monoclonal
Clone: EPR26545-166
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR