Drukuj

Anti-Caveolin-1 Rabbit Polyclonal Antibody

Dostawca: Abcam
AB231448-500 AB231448-100 AB231448-1
ABCAAB231448-500EA 4080 PLN
ABCAAB231448-500 ABCAAB231448-100 ABCAAB231448-1
Anti-Caveolin-1 Rabbit Polyclonal Antibody
Przeciwciała
Rabbit polyclonal to Caveolin-1 - N-terminal.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Caveolin-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR