Drukuj

Anti-PI 3 Kinase p85 alpha Rabbit Monoclonal Antibody [clone: EPR18702]

Dostawca: Abcam
AB191606-100 AB191606-1ML AB191606-20UL
ABCAAB191606-100EA 3420 PLN
ABCAAB191606-100 ABCAAB191606-1ML ABCAAB191606-20UL
Anti-PI 3 Kinase p85 alpha Rabbit Monoclonal Antibody [clone: EPR18702]
Przeciwciała
Rabbit monoclonal [EPR18702] to PI 3 Kinase p85 alpha.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PI 3 Kinase p85 alpha
Clonality: Monoclonal
Clone: EPR18702
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;African green monkey
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR