Drukuj

Anti-Actin Regulatory Protein CAPG/MCP Rabbit Monoclonal Antibody [clone: EPR13194-34]

Dostawca: Abcam
AB181095-100 AB181095-1ML
ABCAAB181095-100EA 3170 PLN
ABCAAB181095-100 ABCAAB181095-1ML
Anti-Actin Regulatory Protein CAPG/MCP Rabbit Monoclonal Antibody [clone: EPR13194-34]
Przeciwciała
Rabbit monoclonal [EPR13194-34] to Actin Regulatory Protein CAPG/MCP.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Actin Regulatory Protein CAPG/MCP
Clonality: Monoclonal
Clone: EPR13194-34
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR