Drukuj

Anti-TCR delta Rabbit Monoclonal Antibody [clone: EPR27043-16]

Dostawca: Abcam
AB313573-100UL AB313573-20UL AB313573-1ML
ABCAAB313573-100ULEA 3090 PLN
ABCAAB313573-100UL ABCAAB313573-20UL ABCAAB313573-1ML
Anti-TCR delta Rabbit Monoclonal Antibody [clone: EPR27043-16]
Przeciwciała
Rabbit monoclonal [EPR27043-16] to TCR delta.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: TCR delta
Clonality: Monoclonal
Clone: EPR27043-16
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR