Drukuj

Anti-RBX1 Rabbit Monoclonal Antibody [clone: EPR20185]

Dostawca: Abcam
AB251569-1 AB251569-100
ABCAAB251569-1EA 40660 PLN
ABCAAB251569-1 ABCAAB251569-100
Anti-RBX1 Rabbit Monoclonal Antibody [clone: EPR20185]
Przeciwciała
Rabbit monoclonal [EPR20185] to RBX1 - BSA and Azide free.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RBX1
Clonality: Monoclonal
Clone: EPR20185
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR