Drukuj

Anti-AUH Rabbit Monoclonal Antibody [clone: EPR11087(B)]

Dostawca: Abcam
AB155980-100 AB155980-1ML
ABCAAB155980-100EA 3140 PLN
ABCAAB155980-100 ABCAAB155980-1ML
Anti-AUH Rabbit Monoclonal Antibody [clone: EPR11087(B)]
Przeciwciała
Rabbit monoclonal [EPR11087(B)] to AUH.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: AUH
Clonality: Monoclonal
Clone: EPR11087(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR