Drukuj

Anti-IFNG Rabbit Monoclonal Antibody [clone: EPR24641-20]

Dostawca: Abcam
AB289983-100 AB289983-1
ABCAAB289983-100EA 4170 PLN
ABCAAB289983-100 ABCAAB289983-1
Anti-IFNG Rabbit Monoclonal Antibody [clone: EPR24641-20]
Przeciwciała
Rabbit monoclonal [EPR24641-20] to Interferon gamma - BSA and Azide free (Detector).

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: EPR24641-20
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR