Drukuj

Anti-Transferrin Receptor Mouse Monoclonal Antibody [clone: 2/Transferrin H1L1]

Dostawca: Abcam
AB300658-100 AB300658-1
ABCAAB300658-100EA 4530 PLN
ABCAAB300658-100 ABCAAB300658-1
Anti-Transferrin Receptor Mouse Monoclonal Antibody [clone: 2/Transferrin H1L1]
Przeciwciała
Mouse monoclonal [2/Transferrin] to Transferrin Receptor - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Transferrin Receptor
Clonality: Monoclonal
Clone: 2/Transferrin H1L1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR