Drukuj

Anti-CD13 Rabbit Monoclonal Antibody [clone: SP182]

Dostawca: Abcam
AB183358-100 AB183358-500 AB183358-1
ABCAAB183358-100EA 3170 PLN
ABCAAB183358-100 ABCAAB183358-500 ABCAAB183358-1
Anti-CD13 Rabbit Monoclonal Antibody [clone: SP182]
Przeciwciała
Rabbit monoclonal [SP182] to CD13.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: SP182
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR