Drukuj

Anti-RNase H1/RNH1 Rabbit Polyclonal Antibody

Dostawca: Abcam
AB229078-100 AB229078-25
ABCAAB229078-100EA 2840 PLN
ABCAAB229078-100 ABCAAB229078-25
Anti-RNase H1/RNH1 Rabbit Polyclonal Antibody
Przeciwciała
Rabbit polyclonal to RNase H1/RNH1.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RNase H1/RNH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Zebrafish
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR