Drukuj

Anti-GSDMA Rabbit Monoclonal Antibody [clone: EPR19858-17]

Dostawca: Abcam
AB214818-100 AB214818-1ML
ABCAAB214818-100EA 3390 PLN
ABCAAB214818-100 ABCAAB214818-1ML
Anti-GSDMA Rabbit Monoclonal Antibody [clone: EPR19858-17]
Przeciwciała
Rabbit monoclonal [EPR19858-17] to GSDMA.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GSDMA
Clonality: Monoclonal
Clone: EPR19858-17
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR