Drukuj

Anti-Eph receptor B1/NET Rabbit Monoclonal Antibody [clone: EPR6457]

Dostawca: Abcam
AB248301-100 AB248301-1
ABCAAB248301-100EA 4530 PLN
ABCAAB248301-100 ABCAAB248301-1
Anti-Eph receptor B1/NET Rabbit Monoclonal Antibody [clone: EPR6457]
Przeciwciała
Rabbit monoclonal [EPR6457] to Eph receptor B1/NET - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Eph receptor B1/NET
Clonality: Monoclonal
Clone: EPR6457
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;African green monkey
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR