Drukuj

Anti-YKL-40/CHI3L1 Rabbit Monoclonal Antibody [clone: EPR27396-56] (Alexa Fluor® 488)

Dostawca: Abcam
AB314164-100UL AB314164-10UL
ABCAAB314164-100ULEA 3640 PLN
ABCAAB314164-100UL ABCAAB314164-10UL
Anti-YKL-40/CHI3L1 Rabbit Monoclonal Antibody [clone: EPR27396-56] (Alexa Fluor® 488)
Przeciwciała
Alexa Fluor® 488 Rabbit monoclonal [EPR27396-56] to YKL-40/CHI3L1.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: YKL-40/CHI3L1
Clonality: Monoclonal
Clone: EPR27396-56
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR