Drukuj

Anti-JLP Rabbit Monoclonal Antibody [clone: EPR25121-32]

Dostawca: Abcam
AB307426-100 AB307426-20UL AB307426-1ML
ABCAAB307426-100EA 3090 PLN
ABCAAB307426-100 ABCAAB307426-20UL ABCAAB307426-1ML
Anti-JLP Rabbit Monoclonal Antibody [clone: EPR25121-32]
Przeciwciała
Rabbit monoclonal [EPR25121-32] to JLP.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: JLP
Clonality: Monoclonal
Clone: EPR25121-32
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR